Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009536.4 | NP_033562.3 | 3 - 4 | 515 | 82 | |
GenBank mRNA | AF483478.1 | - | 3 - 4 | 374 | 82 | |
AF483479.1 | - | 3 - 4 | 374 | 82 | ||
AK075995.1 | - | 3 - 4 | 464 | 82 | ||
AK135831.1 | - | 3 - 4 | 443 | 82 | ||
AK135867.1 | - | 3 - 4 | 443 | 82 | ||
AK169812.1 | - | 3 - 4 | 470 | 82 | ||
AK190793.1 | - | 1 - 2 | 127 | 82 | ||
AK191008.1 | - | 1 - 2 | 104 | 82 | ||
AK192790.1 | - | 1 - 2 | 119 | 82 | ||
AK192964.1 | - | 2 - 3 | 273 | 82 | ||
AK195513.1 | - | 1 - 2 | 119 | 82 | ||
AK218242.1 | - | 1 - 2 | 93 | 82 | ||
BC058686.1 | - | 3 - 4 | 474 | 6411635 | 82 | |
D87663.1 | - | 3 - 4 | 402 | 82 | ||
Z19599.1 | - | 3 - 4 | 457 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |