Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004157.2 | NP_001004157.2 | 2 - 3 | 213 | 70 | |
GenBank mRNA | AK133477.1 | - | 2 - 3 | 196 | 70 | |
AK220215.1 | - | 2 - 3 | 197 | 70 | ||
BC080288.1 | - | 2 - 3 | 213 | 6411836 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |