Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004157.2 | NP_001004157.2 | 4 - 5 | 833 | 75 | |
GenBank mRNA | AK133477.1 | - | 4 - 5 | 816 | 75 | |
AK220215.1 | - | 4 - 5 | 817 | 75 | ||
BC057600.1 | - | 3 - 4 | 740 | 75 | ||
BC080288.1 | - | 4 - 5 | 833 | 6411836 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |