Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013599.3 | NP_038627.1 | 11 - 12 | 1989 | 68 | |
GenBank mRNA | AK004651.1 | - | 11 - 12 | 1970 | 68 | |
AK142787.1 | - | 11 - 12 | 1971 | 68 | ||
AK159292.1 | - | 11 - 12 | 1971 | 68 | ||
AK161176.1 | - | 11 - 12 | 1970 | 68 | ||
BC046991.1 | - | 11 - 12 | 1957 | 6309245 | 68 | |
D12712.1 | - | 11 - 12 | 1978 | 68 | ||
S67830.1 | - | 11 - 12 | 68 | |||
X72795.1 | - | 9 - 10 | 68 | |||
Z27231.1 | - | 11 - 12 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |