Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011125.2 | NP_035255.1 | 1 - 2 | 130 | 70 | |
GenBank mRNA | AK148330.1 | - | 1 - 2 | 135 | 70 | |
AK149730.1 | - | 1 - 2 | 61 | 70 | ||
U37226.1 | - | 1 - 2 | 60 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons |