Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011125.2 | NP_035255.1 | 6 - 7 | 688 | 57 | |
GenBank mRNA | AK148330.1 | - | 6 - 7 | 693 | 57 | |
AK149730.1 | - | 6 - 7 | 619 | 57 | ||
BC003782.1 | - | 6 - 7 | 622 | 3491360 | 57 | |
CT010317.1 | - | 5 - 6 | 552 | 57 | ||
U28960.1 | - | 5 - 6 | 546 | 57 | ||
U37226.1 | - | 6 - 7 | 618 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |