Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038492.2 | NP_001033581.1 | 3 - 4 | 609 | 63 | |
NM_008906.4 | NP_032932.2 | 3 - 4 | 362 | 63 | ||
GenBank mRNA | AK019861.1 | - | 3 - 4 | 567 | 63 | |
AK032197.1 | - | 3 - 4 | 329 | 63 | ||
AK159150.1 | - | 3 - 4 | 330 | 63 | ||
AK159766.1 | - | 3 - 4 | 325 | 63 | ||
BC018534.1 | - | 3 - 4 | 318 | 2651479 | 63 | |
J05261.1 | - | 3 - 4 | 497 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |