Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310786 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310644 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310784 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310642 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310783 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310788 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310785 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310787 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539656 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
677103 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404307 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
935187 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
789022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
AK038739.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
AK081422.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
AK165904.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
BC113205.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
L12460.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
889958 |
NM_008926.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607043 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726158 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150395 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
108206 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169491 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310784 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310788 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310786 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310800 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310783 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310787 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310798 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310791 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310793 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310785 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27310803 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02177178 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169491 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02177192 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK149699.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK082158.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
NM_133993.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P207403 |
AK009972.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122788 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169491 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08873696 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08873703 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310644 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310784 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310788 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310642 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310787 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310786 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310785 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27310783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310894 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310913 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310960 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310877 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27310872 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310784 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310828 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310802 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310851 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310798 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08398260 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310832 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08398262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310809 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310796 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08398272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310788 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310785 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08752985 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08359137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08679785 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310821 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310794 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310800 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310791 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27310810 |
- |
- |