Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133967.3 | NP_598728.1 | 6 - 7 | 1101 | 96 | |
GenBank mRNA | AK041905.1 | - | 6 - 7 | 970 | 96 | |
AK090333.1 | - | 6 - 7 | 947 | 96 | ||
AK157968.1 | - | 6 - 7 | 954 | 96 | ||
AK170639.1 | - | 6 - 7 | 954 | 96 | ||
AK170923.1 | - | 6 - 7 | 924 | 96 | ||
AK172462.1 | - | 6 - 7 | 935 | 96 | ||
AK213858.1 | - | 3 - 4 | 229 | 96 | ||
AY166673.1 | - | 4 - 5 | 619 | 96 | ||
BC013467.1 | - | 5 - 6 | 652 | 4218025 | 96 | |
BC029666.1 | - | 1 - 2 | 55 | 96 | ||
BC075666.1 | - | 6 - 7 | 1101 | 30605163 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |