Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009462.1 | NP_033488.1 | 11 - 12 | 2046 | 70 | |
GenBank mRNA | AK077453.1 | - | 11 - 12 | 2068 | 70 | |
AK129083.1 | - | 11 - 12 | 2054 | 70 | ||
AK153675.1 | - | 11 - 12 | 2538 | 70 | ||
AK157970.1 | - | 11 - 12 | 2070 | 70 | ||
AK165156.1 | - | 11 - 12 | 2066 | 70 | ||
AK166243.1 | - | 11 - 12 | 2062 | 70 | ||
AK172443.1 | - | 11 - 12 | 2068 | 70 | ||
AK202299.1 | - | 2 - 3 | 229 | 70 | ||
BC007134.1 | - | 11 - 12 | 2048 | 3491297 | 70 | |
D84096.1 | - | 11 - 12 | 2046 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |