Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291225.1 | NP_001278154.1 | 2 - 3 | 937 | 110 | |
NM_021286.4 | NP_067261.2 | 2 - 3 | 937 | 110 | ||
GenBank mRNA | AK032277.1 | - | 2 - 3 | 860 | 110 | |
AK147705.1 | - | 2 - 3 | 856 | 110 | ||
AK165757.1 | - | 2 - 3 | 721 | 110 | ||
BC053011.1 | - | 2 - 3 | 833 | 5698186 | 110 | |
BC055345.1 | - | 2 - 3 | 937 | 6833344 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |