Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153145.4 | NP_694785.3 | 24 - 25 | 3387 | 120 | |
GenBank mRNA | AB255881.1 | - | 23 - 24 | 3306 | 120 | |
AB255882.1 | - | 24 - 25 | 3394 | 120 | ||
AF498360.1 | - | 23 - 24 | 3315 | 120 | ||
AK031843.1 | - | 5 - 6 | 618 | 120 | ||
AY732492.1 | - | 23 - 24 | 3315 | 120 | ||
BC026496.1 | - | 4 - 5 | 512 | 120 | ||
BC060032.1 | - | 6 - 7 | 881 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |