Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004144.1 | NP_001004144.1 | 14 - 15 | 1807 | 55 | |
GenBank mRNA | AK050701.1 | - | 1 - 2 | 125 | 55 | |
BC079870.1 | - | 14 - 15 | 1807 | 6831910 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |