Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166506.1 | NP_001159978.1 | 5 - 6 | 686 | 155 | |
NM_001166507.1 | NP_001159979.1 | 5 - 6 | 686 | 155 | ||
NM_028777.3 | NP_083053.2 | 5 - 6 | 686 | 155 | ||
GenBank mRNA | AK004818.1 | - | 5 - 6 | 611 | 155 | |
AK047396.1 | - | 5 - 6 | 691 | 155 | ||
AK172947.1 | - | 2 - 3 | 221 | 155 | ||
BC005766.1 | - | 5 - 6 | 644 | 2649054 | 155 | |
BC086620.1 | - | 5 - 6 | 620 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |