Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144892.1 | NP_659141.1 | 2 - 3 | 165 | 91 | |
GenBank mRNA | BC017152.1 | - | 2 - 3 | 165 | 4511696 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |