Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133763.1 | NP_598524.1 | 10 - 11 | 763 | 75 | |
GenBank mRNA | AB045974.1 | - | 10 - 11 | 756 | 75 | |
AK145239.1 | - | 10 - 11 | 789 | 75 | ||
AK168357.1 | - | 10 - 11 | 783 | 75 | ||
AK185487.1 | - | 4 - 5 | 190 | 75 | ||
AK187737.1 | - | 4 - 5 | 189 | 75 | ||
BC003486.1 | - | 10 - 11 | 763 | 2811159 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |