Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166685.1 | NP_001160157.1 | 10 - 11 | 1275 | 90 | |
NM_177088.3 | NP_796062.2 | 11 - 12 | 1404 | 90 | ||
GenBank mRNA | AK052705.1 | - | 11 - 12 | 1332 | 90 | |
AK076400.1 | - | 11 - 12 | 1382 | 90 | ||
BC090675.1 | - | 11 - 12 | 1360 | 90 | ||
BC116308.1 | - | 10 - 11 | 1241 | 40095242 | 90 | |
BC116309.1 | - | 10 - 11 | 1254 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |