Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166685.1 | NP_001160157.1 | 8 - 9 | 994 | 100 | |
NM_177088.3 | NP_796062.2 | 9 - 10 | 1123 | 100 | ||
GenBank mRNA | AK052705.1 | - | 9 - 10 | 1051 | 100 | |
AK076400.1 | - | 9 - 10 | 1101 | 100 | ||
BC090675.1 | - | 9 - 10 | 1079 | 100 | ||
BC116308.1 | - | 8 - 9 | 960 | 40095242 | 100 | |
BC116309.1 | - | 8 - 9 | 973 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |