Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172566.4 | NP_766154.2 | 3 - 4 | 885 | 81 | |
GenBank mRNA | AK086362.1 | - | 3 - 4 | 886 | 81 | |
AK157156.1 | - | 3 - 4 | 879 | 81 | ||
BC119592.1 | - | 3 - 4 | 886 | 40059040 | 81 | |
BC119593.1 | - | 3 - 4 | 886 | 40059041 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |