Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037723.3 | NP_001032812.2 | 12 - 13 | 1963 | 112 | |
NM_001037724.4 | NP_001032813.1 | 12 - 13 | 1899 | 112 | ||
NM_001109756.1 | NP_001103226.1 | 12 - 13 | 1867 | 112 | ||
NM_007406.2 | NP_031432.2 | 12 - 13 | 2051 | 112 | ||
GenBank mRNA | AK138911.1 | - | 12 - 13 | 1883 | 112 | |
AK154262.1 | - | 12 - 13 | 1954 | 112 | ||
AK154652.1 | - | 12 - 13 | 1869 | 112 | ||
AK155646.1 | - | 12 - 13 | 1868 | 112 | ||
AK155831.1 | - | 12 - 13 | 1870 | 112 | ||
BC057682.1 | - | 4 - 5 | 475 | 112 | ||
BC115833.1 | - | 10 - 11 | 1745 | 40086640 | 112 | |
U12919.1 | - | 12 - 13 | 2052 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |