Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133779.2 | NP_598540.2 | 5 - 6 | 734 | 58 | |
GenBank mRNA | AK019717.2 | - | 5 - 6 | 725 | 58 | |
AK044499.1 | - | 5 - 6 | 718 | 58 | ||
AK049171.1 | - | 5 - 6 | 728 | 58 | ||
AK049990.1 | - | 5 - 6 | 727 | 58 | ||
AK090020.1 | - | 5 - 6 | 726 | 58 | ||
AK155057.1 | - | 5 - 6 | 721 | 58 | ||
AK157226.1 | - | 5 - 6 | 714 | 58 | ||
AK162573.1 | - | 5 - 6 | 714 | 58 | ||
BC016520.1 | - | 6 - 7 | 788 | 58 | ||
BC034175.1 | - | 5 - 6 | 710 | 5026752 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |