Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282000.1 | NP_001268929.1 | 19 - 20 | 3069 | 77 | |
NM_001282001.1 | NP_001268930.1 | 20 - 21 | 3168 | 77 | ||
NM_011250.4 | NP_035380.3 | 20 - 21 | 3198 | 77 | ||
GenBank mRNA | AK160027.1 | - | 20 - 21 | 3145 | 77 | |
BC020528.1 | - | 20 - 21 | 4240592 | 77 | ||
U36799.1 | - | 20 - 21 | 3068 | 77 | ||
U47333.1 | - | 19 - 20 | 3011 | 77 | ||
U50850.1 | - | 20 - 21 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |