Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080855.2 | NP_543131.1 | 2 - 3 | 332 | 67 | |
GenBank mRNA | AB030244.1 | - | 2 - 3 | 343 | 67 | |
BC132449.1 | - | 2 - 3 | 330 | 40130726 | 67 | |
BC137853.1 | - | 2 - 3 | 330 | 8860872 | 67 | |
CJ102007.1 | - | 2 - 3 | 175 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |