Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003951.2 | NP_001003951.1 | 9 - 10 | 1127 | 86 | |
GenBank mRNA | AB186393.1 | - | 9 - 10 | 1127 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |