Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smurf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025481.2 | NP_079757.2 | 7 - 8 | 1055 | 151 | |
GenBank mRNA | AK012060.1 | - | 6 - 7 | 1018 | 151 | |
AY685230.1 | - | 7 - 8 | 735 | 151 | ||
BC138786.1 | - | 8 - 9 | 620 | 8861808 | 151 | |
BC138788.1 | - | 8 - 9 | 631 | 8861810 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |