Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 19 - 20 | 3699 | 106 | |
GenBank mRNA | AK087907.1 | - | 4 - 5 | 612 | 106 | |
AK220342.1 | - | 11 - 12 | 1261 | 106 | ||
BC068235.1 | - | 8 - 9 | 1768 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |