Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026922.1 | NP_081198.1 | 5 - 6 | 598 | 111 | |
GenBank mRNA | AK137687.1 | - | 5 - 6 | 599 | 111 | |
AK220351.1 | - | 3 - 4 | 183 | 111 | ||
EF688288.1 | - | 5 - 6 | 442 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Transporter Detect Genomic DNA Mus musculus Probe spans exons |