Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ftsj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025310.2 | NP_079586.1 | 6 - 7 | 471 | 60 | |
GenBank mRNA | AK004996.2 | - | 6 - 7 | 471 | 60 | |
AK031514.1 | - | 6 - 7 | 458 | 60 | ||
AK145490.1 | - | 6 - 7 | 496 | 60 | ||
BC012281.1 | - | 6 - 7 | 468 | 4012961 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |