Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK050233.1 | - | 1 - 2 | 121 | 161 | |
BC099926.1 | - | 1 - 2 | 139 | 161 | ||
BC144819.1 | - | 1 - 2 | 141 | 161 | ||
BC145801.1 | - | 1 - 2 | 141 | 40131104 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |