Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900026302.mm.1 |
NM_177771.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026302.mm.1 |
NM_177771.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026302.mm.1 |
TC0900003112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900026300.mm.1 |
NM_177771.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026300.mm.1 |
NM_177771.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026300.mm.1 |
TC0900003112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900026303.mm.1 |
NM_177771.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026303.mm.1 |
NM_177771.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026303.mm.1 |
TC0900003112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900026299.mm.1 |
NM_177771.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026299.mm.1 |
NM_177771.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026299.mm.1 |
TC0900003112.mm.1 |
MTA TC |