Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009569.3 | NP_033595.1 | 6 - 7 | 1101 | 59 | |
GenBank mRNA | AF006492.1 | - | 6 - 7 | 1105 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |