Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029993.1 | NP_001025164.1 | 2 - 3 | 786 | 81 | |
NM_001029994.1 | NP_001025165.1 | 2 - 3 | 786 | 81 | ||
GenBank mRNA | AK017933.1 | - | 2 - 3 | 763 | 81 | |
AK031413.1 | - | 2 - 3 | 735 | 81 | ||
AK159881.1 | - | 2 - 3 | 722 | 81 | ||
AK165989.1 | - | 1 - 2 | 137 | 81 | ||
AK166802.1 | - | 2 - 3 | 741 | 81 | ||
BC030495.1 | - | 2 - 3 | 766 | 5400494 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |