Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029993.1 | NP_001025164.1 | 3 - 4 | 876 | 65 | |
GenBank mRNA | AK017933.1 | - | 3 - 4 | 853 | 65 | |
AK031413.1 | - | 3 - 4 | 825 | 65 | ||
AK159881.1 | - | 3 - 4 | 812 | 65 | ||
AK166802.1 | - | 3 - 4 | 831 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |