Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002764.1 | NP_001002764.1 | 16 - 17 | 3874 | 85 | |
GenBank mRNA | AK129203.1 | - | 15 - 16 | 2493 | 85 | |
BC037179.1 | - | 14 - 15 | 2164 | 85 | ||
BC047279.1 | - | 16 - 17 | 3886 | 85 | ||
BC066040.1 | - | 16 - 17 | 3874 | 6410245 | 85 | |
BC077715.1 | - | 16 - 17 | 3837 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |