Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199283.1 | NP_001186212.1 | 5 - 6 | 844 | 57 | |
NM_001199284.1 | NP_001186213.1 | 5 - 6 | 626 | 57 | ||
NM_173388.2 | NP_775564.1 | 5 - 6 | 589 | 57 | ||
GenBank mRNA | AB120363.1 | - | 5 - 6 | 622 | 57 | |
AK143773.1 | - | 5 - 6 | 687 | 57 | ||
AK159144.1 | - | 5 - 6 | 812 | 57 | ||
AK172685.1 | - | 5 - 6 | 614 | 57 | ||
BC042513.1 | - | 5 - 6 | 585 | 3964020 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |