Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002929.4 | NP_001002929.3 | 7 - 8 | 684 | 59 | |
GenBank mRNA | AF498263.1 | - | 7 - 8 | 598 | 59 | |
AK017632.1 | - | 7 - 8 | 649 | 59 | ||
BC079856.1 | - | 7 - 8 | 659 | 6403446 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |