Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178798.3 | NP_848913.1 | 2 - 3 | 633 | 85 | |
GenBank mRNA | AK030762.1 | - | 3 - 4 | 435 | 85 | |
AK046548.1 | - | 2 - 3 | 633 | 85 | ||
AK049478.1 | - | 2 - 3 | 633 | 85 | ||
AK053899.1 | - | 2 - 3 | 642 | 85 | ||
AK088592.1 | - | 2 - 3 | 599 | 85 | ||
AK157127.1 | - | 2 - 3 | 772 | 85 | ||
AK157379.1 | - | 2 - 3 | 634 | 85 | ||
AK164666.1 | - | 2 - 3 | 634 | 85 | ||
AK165629.1 | - | 2 - 3 | 650 | 85 | ||
AK165979.1 | - | 2 - 3 | 652 | 85 | ||
AK202042.1 | - | 1 - 2 | 352 | 85 | ||
BC038404.1 | - | 1 - 2 | 567 | 4949962 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |