Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198308.1 | NP_938050.1 | 10 - 11 | 1443 | 92 | |
GenBank mRNA | AK029575.1 | - | 10 - 11 | 1534 | 92 | |
AK078515.1 | - | 4 - 5 | 534 | 92 | ||
AY223867.1 | - | 10 - 11 | 1443 | 92 | ||
BC120745.1 | - | 9 - 10 | 1373 | 40129668 | 92 | |
BC125425.1 | - | 9 - 10 | 1373 | 40129940 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |