Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170742.1 | NP_001164213.1 | 20 - 21 | 2203 | 75 | |
NM_001170743.1 | NP_001164214.1 | 21 - 22 | 2260 | 75 | ||
NM_176923.4 | NP_795897.2 | 20 - 21 | 2203 | 75 | ||
GenBank mRNA | AK030644.1 | - | 20 - 21 | 2084 | 75 | |
AK040807.1 | - | 5 - 6 | 490 | 75 | ||
AK136303.1 | - | 16 - 17 | 1755 | 75 | ||
AK170910.1 | - | 20 - 21 | 2202 | 75 | ||
AK173301.1 | - | 21 - 22 | 2223 | 75 | ||
BC066082.1 | - | 20 - 21 | 2188 | 30355918 | 75 | |
EF011622.1 | - | 20 - 21 | 2188 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |