Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252483.1 | NP_001239412.1 | 1 - 2 | 167 | 61 | |
GenBank mRNA | AY601911.1 | - | 1 - 2 | 167 | 61 | |
BC115572.1 | - | 1 - 2 | 66 | 40102153 | 61 | |
BC115573.1 | - | 1 - 2 | 66 | 40102154 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |