Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291076.1 | NP_001278005.1 | 2 - 3 | 246 | 146 | |
NM_001291077.1 | NP_001278006.1 | 2 - 3 | 246 | 146 | ||
NM_198214.3 | NP_937857.1 | 2 - 3 | 246 | 146 | ||
GenBank mRNA | AK032279.1 | - | 2 - 3 | 187 | 146 | |
AK122262.1 | - | 2 - 3 | 186 | 146 | ||
BC072567.1 | - | 2 - 3 | 209 | 146 | ||
BC096541.1 | - | 2 - 3 | 138 | 146 | ||
BC117896.1 | - | 2 - 3 | 172 | 40096951 | 146 | |
BC117897.1 | - | 2 - 3 | 172 | 40096952 | 146 | |
BC139792.1 | - | 2 - 3 | 172 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |