Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291076.1 | NP_001278005.1 | 3 - 4 | 344 | 90 | |
GenBank mRNA | AK032279.1 | - | 3 - 4 | 285 | 90 | |
BC117896.1 | - | 3 - 4 | 270 | 40096951 | 90 | |
BC139792.1 | - | 3 - 4 | 270 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |