Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009808.4 | NP_033938.3 | 3 - 4 | 356 | 132 | |
GenBank mRNA | AK030309.1 | - | 3 - 4 | 331 | 132 | |
AK046681.1 | - | 3 - 4 | 355 | 132 | ||
AK053407.1 | - | 3 - 4 | 336 | 132 | ||
AK077256.1 | - | 3 - 4 | 358 | 132 | ||
AK080045.1 | - | 3 - 4 | 357 | 132 | ||
AK161525.1 | - | 3 - 4 | 342 | 132 | ||
BC028979.1 | - | 3 - 4 | 343 | 5375109 | 132 | |
Y13090.1 | - | 2 - 3 | 346 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |