Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172758.4 | NP_766346.1 | 10 - 11 | 1479 | 68 | |
GenBank mRNA | AK052548.1 | - | 10 - 11 | 1480 | 68 | |
AK171137.1 | - | 10 - 11 | 1487 | 68 | ||
BC031853.1 | - | 8 - 9 | 1354 | 4009949 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |