Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172758.4 | NP_766346.1 | 7 - 8 | 1015 | 60 | |
GenBank mRNA | AK052548.1 | - | 7 - 8 | 1016 | 60 | |
AK171137.1 | - | 7 - 8 | 1023 | 60 | ||
AK194183.1 | - | 2 - 3 | 131 | 60 | ||
BC031853.1 | - | 5 - 6 | 890 | 4009949 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |