Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133973.2 | NP_598734.1 | 1 - 2 | 190 | 68 | |
GenBank mRNA | AK156466.1 | - | 1 - 2 | 181 | 68 | |
AK172465.1 | - | 1 - 2 | 181 | 68 | ||
BC023120.1 | - | 1 - 2 | 167 | 4161858 | 68 | |
CN843612.1 | - | 1 - 2 | 209 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |