Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133973.2 | NP_598734.1 | 4 - 5 | 556 | 64 | |
GenBank mRNA | AK156466.1 | - | 4 - 5 | 547 | 64 | |
AK184101.1 | - | 1 - 2 | 170 | 64 | ||
AK203917.1 | - | 2 - 3 | 209 | 64 | ||
AY223866.1 | - | 4 - 5 | 583 | 64 | ||
BC023120.1 | - | 4 - 5 | 533 | 4161858 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |