Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133953.2 | NP_598714.1 | 9 - 10 | 1430 | 91 | |
GenBank mRNA | AK040625.1 | - | 9 - 10 | 1404 | 91 | |
AK046845.1 | - | 9 - 10 | 1415 | 91 | ||
AK088268.1 | - | 9 - 10 | 1403 | 91 | ||
AK128988.1 | - | 9 - 10 | 1420 | 91 | ||
AK129035.1 | - | 9 - 10 | 1398 | 91 | ||
AK147914.1 | - | 9 - 10 | 1421 | 91 | ||
AK164199.1 | - | 9 - 10 | 1421 | 91 | ||
AK203671.1 | - | 2 - 3 | 261 | 91 | ||
BC011412.1 | - | 9 - 10 | 1411 | 3600115 | 91 | |
BC042580.1 | - | 9 - 10 | 1357 | 5367372 | 91 | |
BC139015.1 | - | 8 - 9 | 1294 | 8862037 | 91 | |
BC139016.1 | - | 8 - 9 | 1294 | 8862038 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |