Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800010858.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010858.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010858.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010860.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010860.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010860.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010851.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010851.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010851.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010863.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010863.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010863.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010859.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010859.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010859.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010852.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010852.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010852.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010862.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010862.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010862.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010854.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010854.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010854.mm.1 |
TC0800001324.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010861.mm.1 |
NM_009179.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010861.mm.1 |
NM_009179.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010861.mm.1 |
TC0800001324.mm.1 |
MTA TC |