Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 2 - 3 | 996 | 89 | |
GenBank mRNA | AK032416.1 | - | 2 - 3 | 959 | 89 | |
AK076799.1 | - | 2 - 3 | 569 | 89 | ||
AK141443.1 | - | 2 - 3 | 546 | 89 | ||
AK151018.1 | - | 2 - 3 | 966 | 89 | ||
AK152024.1 | - | 2 - 3 | 966 | 89 | ||
AK172567.1 | - | 2 - 3 | 962 | 89 | ||
AK173313.1 | - | 2 - 3 | 615 | 89 | ||
BC010833.1 | - | 2 - 3 | 611 | 4235165 | 89 | |
BC054121.1 | - | 2 - 3 | 982 | 6393910 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |